Learn More
Trajan Scientific Tubing PEEK Natural Medical 1/EA
SDP
Catalog No. 5000494294
Encompass_Preferred
1/16 OD x 0 040 ID x 100 m length PEEK tubing natural
Catalog No. 50-004-94294 Supplier Trajan Scientific Supplier No. 1302040000100M
1/16 OD x 0 040 ID x 100 m length PEEK tubing natural