Learn More
Trajan Scientific Tubing PEEK Natural Medical 1/EA
SDP
Catalog No. 5000494164
Encompass_Preferred
1/16 OD x 0 0025 ID x 100 m length PEEK tubing natural
Catalog No. 50-004-94164 Supplier Trajan Scientific Supplier No. 1302002500100M
1/16 OD x 0 0025 ID x 100 m length PEEK tubing natural