Learn More
Trajan Scientific Tubing PEEK Natural Medical 1/EA
SDP
Catalog No. 5000494210
Encompass_Preferred
1/8 OD x 0 080 ID x 100 length PEEK tubing natural
Catalog No. 50-004-94210 Supplier Trajan Scientific Supplier No. 1303080000100F
1/8 OD x 0 080 ID x 100 length PEEK tubing natural