Learn More
Microsolv Technology Corporation Tubing, PEEK, 0.0025" ID, 1/16" OD, Super-T Grade, Natural Color. 5 Meter Roll. 1 EA.
Catalog No. 502271468
Encompass_Preferred
This Super-T™ grade ( Premium ), PEEK tubing is extruded from virgin PEEK ( Victrex ) material without adding any additives. The tubing is a natural PEEK which eliminates leachables due to color. The internal and outer dimensional tolerances of this tubing is +/- 0.025mm. Color Code: Natural, ( No Stripe ) for 0.0025" ( 0.064mm ) ID. Pressure rating: 6300 psi.
This item is not returnable. View return policy
Catalog No. 50-227-1468 Supplier Microsolv Technology Corporation Supplier No. 47305ST02
This Super-T™ grade ( Premium ), PEEK tubing is extruded from virgin PEEK ( Victrex ) material without adding any additives. The tubing is a natural PEEK which eliminates leachables due to color. The internal and outer dimensional tolerances of this tubing is +/- 0.025mm. Color Code: Natural, ( No Stripe ) for 0.0025" ( 0.064mm ) ID. Pressure rating: 6300 psi.