Learn More
Trajan Scientific Tubing PEEK Orange Medical 1/EA
SDP
Catalog No. 5000493993
Encompass_Preferred
1/16 OD x 0 020 ID x 100 m length PEEK tubing orange
Catalog No. 50-004-93993 Supplier Trajan Scientific Supplier No. 1302020004100M
1/16 OD x 0 020 ID x 100 m length PEEK tubing orange