Learn More
Trajan Scientific Tubing PEEK Orange Medical 1/EA
SDP
Supplier:  Trajan Scientific 130201300450F
Encompass_Preferred
1/16 OD x 0 013 ID x 50 length PEEK tubing orange
Catalog No. 50-004-93626