Learn More
Trajan Scientific Tubing PEEK Orange Medical 1/EA
SDP
Supplier:  Trajan Scientific 1301020004100F
Encompass_Preferred
1/32 OD x 0 020 ID x 100 length PEEK tubing orange
Catalog No. 50-004-94935