Learn More
Microsolv Technology Corporation Tubing, PEEK, Pre-Cut, 0.010" ID, 1/16th" OD, Super-T Grade, Blue Stripe, 10cm Long. 5 PK.
Catalog No. 502269759
Encompass_Preferred
Properly cutting tubing is one of the most difficult tasks in a chromatography laboratory. These Pre-Cut PEEK Tubes are the perfect solution. This Super-T™ grade ( Premium ), PEEK tubing is extruded from virgin PEEK ( Victrex ) material without adding any additives. The broken yellow stripes are added only to the outside of the tubing, leaving the interior all natural PEEK which eliminates leachables due to color. The internal and outer dimensional tolerances of this tubing is +/- 0.025mm. Color Code: Blue, Stripes for 0.010" ( 0.25mm ) ID. Pressure rating: 5600 psi.
This item is not returnable. View return policy
Catalog No. 50-226-9759 Supplier Microsolv Technology Corporation Supplier No. 473052510
Properly cutting tubing is one of the most difficult tasks in a chromatography laboratory. These Pre-Cut PEEK Tubes are the perfect solution. This Super-T™ grade ( Premium ), PEEK tubing is extruded from virgin PEEK ( Victrex ) material without adding any additives. The broken yellow stripes are added only to the outside of the tubing, leaving the interior all natural PEEK which eliminates leachables due to color. The internal and outer dimensional tolerances of this tubing is +/- 0.025mm. Color Code: Blue, Stripes for 0.010" ( 0.25mm ) ID. Pressure rating: 5600 psi.