Learn More
Trajan Scientific Tubing PEEK Red Medical 1/EA
SDP
Catalog No. 5000494290
Encompass_Preferred
360 m OD x 100 m (0 004 ) ID x 100 length PEEK tubing red
Catalog No. 50-004-94290 Supplier Trajan Scientific Supplier No. 1304004001100F
360 m OD x 100 m (0 004 ) ID x 100 length PEEK tubing red