Learn More
Microsolv Technology Corporation Tubing, PEEK, 0.007" ID, 1/16" OD, HPLC grade, yellow stripe. 25-meter roll. 1 EA.
Catalog No. 502271458
Encompass_Preferred
This HPLC Grade, PEEK tubing is extruded from virgin PEEK ( Victrex ) material without adding any additives. The yellow stripe indicating size is applied to the outside of the tubing leaving the interior all-natural PEEK which eliminates color leachables. The internal and outer dimensional tolerances of this tubing is +/- 0.05mm. Color Code: Yellow Stripe for 0.007" ( 0.18mm ) ID. Pressure rating: 5800 psi with proper mobile phase.
This item is not returnable. View return policy
Catalog No. 50-227-1458 Supplier Microsolv Technology Corporation Supplier No. 4730507C
This HPLC Grade, PEEK tubing is extruded from virgin PEEK ( Victrex ) material without adding any additives. The yellow stripe indicating size is applied to the outside of the tubing leaving the interior all-natural PEEK which eliminates color leachables. The internal and outer dimensional tolerances of this tubing is +/- 0.05mm. Color Code: Yellow Stripe for 0.007" ( 0.18mm ) ID. Pressure rating: 5800 psi with proper mobile phase.