Learn More
Trajan Scientific Tubing PEEK Yellow Medical 1/EA
SDP
Supplier:  Trajan Scientific 1301007002100F
Encompass_Preferred
1/32 OD x 0 007 ID x 100 length PEEK tubing yellow
Catalog No. 50-004-94577