Learn More
Trajan Scientific Tubing PEEKSil Beige Medical 2/PK
SDP
Catalog No. 5000494616
Encompass_Preferred
360 m OD x 0 05 mm ID x 100 mm length PEEKsil tubing beige PK2
Catalog No. 50-004-94616 Supplier Trajan Scientific Supplier No. 14030201000202
360 m OD x 0 05 mm ID x 100 mm length PEEKsil tubing beige PK2