Learn More
Trajan Scientific Tubing PEEKSil Beige Medical 2/PK
SDP
Catalog No. 5000493870
Encompass_Preferred
1/32 OD x 0 05 mm ID x 350 mm length PEEKsil tubing beige PK2
Catalog No. 50-004-93870 Supplier Trajan Scientific Supplier No. 14020203500202
1/32 OD x 0 05 mm ID x 350 mm length PEEKsil tubing beige PK2