Learn More
Trajan Scientific Tubing PEEKSil Beige Medical 2/PK
SDP
Catalog No. 5000493791
Encompass_Preferred
1/16 OD x 0 53 mm ID x 700 mm length PEEKsil tubing beige PK2
Catalog No. 50-004-93791 Supplier Trajan Scientific Supplier No. 14010907000202
1/16 OD x 0 53 mm ID x 700 mm length PEEKsil tubing beige PK2