Learn More
Trajan Scientific Tubing PEEKSil Beige Medical 2/PK
SDP
Catalog No. 5000493902
Encompass_Preferred
1/16 OD x 0 53 mm ID x 350 mm length PEEKsil tubing beige PK2
Catalog No. 50-004-93902 Supplier Trajan Scientific Supplier No. 14010903500202
1/16 OD x 0 53 mm ID x 350 mm length PEEKsil tubing beige PK2