Learn More
Trajan Scientific Tubing PEEKSil Beige Medical 2/PK
SDP
Catalog No. 5000493817
Encompass_Preferred
360 m OD x 0 05 mm ID x 250 mm length PEEKsil tubing beige PK2
Catalog No. 50-004-93817 Supplier Trajan Scientific Supplier No. 14030202500202
360 m OD x 0 05 mm ID x 250 mm length PEEKsil tubing beige PK2