Learn More
Trajan Scientific Tubing PEEKSil Beige Medical 2/PK
SDP
Catalog No. 5000493749
Encompass_Preferred
1/16 OD x 0 53 mm ID x 500 mm length PEEKsil tubing beige PK2
Catalog No. 50-004-93749 Supplier Trajan Scientific Supplier No. 14010905000202
1/16 OD x 0 53 mm ID x 500 mm length PEEKsil tubing beige PK2