Learn More
Trajan Scientific Tubing PEEKSil Beige Medical 2/PK
SDP
Catalog No. 5000493926
Encompass_Preferred
1/32 OD x 0 05 mm ID x 50 mm length PEEKsil tubing beige PK2
Catalog No. 50-004-93926 Supplier Trajan Scientific Supplier No. 14020200500202
1/32 OD x 0 05 mm ID x 50 mm length PEEKsil tubing beige PK2