Learn More
Trajan Scientific Tubing PEEKSil Beige Medical 2/PK
SDP
Catalog No. 5000494566
Encompass_Preferred
1/16 OD x 0 05 mm ID x 450 mm length PEEKsil tubing beige PK2
Catalog No. 50-004-94566 Supplier Trajan Scientific Supplier No. 14010204500202
1/16 OD x 0 05 mm ID x 450 mm length PEEKsil tubing beige PK2