Learn More
Trajan Scientific Tubing PEEKSil Beige Medical 2/PK
SDP
Catalog No. 5000494044
Encompass_Preferred
360 m OD x 0 05 mm ID x 450 mm length PEEKsil tubing beige PK2
Catalog No. 50-004-94044 Supplier Trajan Scientific Supplier No. 14030204500202
360 m OD x 0 05 mm ID x 450 mm length PEEKsil tubing beige PK2