Learn More
Trajan Scientific Tubing PEEKSil Black Medical 2/PK
SDP
Catalog No. 5000493750
Encompass_Preferred
1/32 OD x 0 075 mm ID x 700 mm length PEEKsil tubing black PK2
Catalog No. 50-004-93750 Supplier Trajan Scientific Supplier No. 14020307000302
Add to Cart
Add to Cart
1/32 OD x 0 075 mm ID x 700 mm length PEEKsil tubing black PK2