Learn More
Trajan Scientific Tubing PEEKSil Black Medical 2/PK
SDP
Catalog No. 5000493679
Encompass_Preferred
1/32 OD x 0 075 mm ID x 500 mm length PEEKsil tubing black PK2
Catalog No. 50-004-93679 Supplier Trajan Scientific Supplier No. 14020305000302
1/32 OD x 0 075 mm ID x 500 mm length PEEKsil tubing black PK2