Learn More
Trajan Scientific Tubing PEEKSil Black Medical 2/PK
SDP
Catalog No. 5000493938
Encompass_Preferred
1/32 OD x 0 075 mm ID x 350 mm length PEEKsil tubing black PK2
Catalog No. 50-004-93938 Supplier Trajan Scientific Supplier No. 14020303500302
1/32 OD x 0 075 mm ID x 350 mm length PEEKsil tubing black PK2