Learn More
Trajan Scientific Tubing PEEKSil Black Medical 2/PK
SDP
Catalog No. 5000494066
Encompass_Preferred
1/32 OD x 0 075 mm ID x 250 mm length PEEKsil tubing black PK2
Catalog No. 50-004-94066 Supplier Trajan Scientific Supplier No. 14020302500302
1/32 OD x 0 075 mm ID x 250 mm length PEEKsil tubing black PK2