Learn More
Trajan Scientific Tubing PEEKSil Black Medical 2/PK
SDP
Catalog No. 5000494374
Encompass_Preferred
1/16 OD x 0 075 mm ID x 200 mm length PEEKsil tubing black PK2
Catalog No. 50-004-94374 Supplier Trajan Scientific Supplier No. 14010302000302
1/16 OD x 0 075 mm ID x 200 mm length PEEKsil tubing black PK2