Learn More
Trajan Scientific Tubing PEEKSil Blue Medical 2/PK
SDP
Catalog No. 5000493890
Encompass_Preferred
1/16 OD x 0 2 mm ID x 550 mm length PEEKsil tubing blue PK2
Catalog No. 50-004-93890 Supplier Trajan Scientific Supplier No. 14010705500702
1/16 OD x 0 2 mm ID x 550 mm length PEEKsil tubing blue PK2