Learn More
Trajan Scientific Tubing PEEKSil Blue Medical 2/PK
SDP
Catalog No. 5000493717
Encompass_Preferred
1/16 OD x 0 2 mm ID x 100 mm length PEEKsil tubing blue PK2
Catalog No. 50-004-93717 Supplier Trajan Scientific Supplier No. 14010701000702
1/16 OD x 0 2 mm ID x 100 mm length PEEKsil tubing blue PK2