Learn More
Trajan Scientific Tubing PEEKSil Blue Medical 2/PK
SDP
Catalog No. 5000493918
Encompass_Preferred
1/16 OD x 0 2 mm ID x 300 mm length PEEKsil tubing blue PK2
Catalog No. 50-004-93918 Supplier Trajan Scientific Supplier No. 14010703000702
1/16 OD x 0 2 mm ID x 300 mm length PEEKsil tubing blue PK2