Learn More
Trajan Scientific Tubing PEEKSil Blue Medical 2/PK
SDP
Catalog No. 5000493861
Encompass_Preferred
1/16 OD x 0 2 mm ID x 650 mm length PEEKsil tubing blue PK2
Catalog No. 50-004-93861 Supplier Trajan Scientific Supplier No. 14010706500702
1/16 OD x 0 2 mm ID x 650 mm length PEEKsil tubing blue PK2