Learn More
Trajan Scientific Tubing PEEKSil Green Medical 2/PK
SDP
Catalog No. 5000493901
Encompass_Preferred
1/16 OD x 0 125 mm ID x 400 mm length PEEKsil tubing green PK2
Catalog No. 50-004-93901 Supplier Trajan Scientific Supplier No. 14011004000902
1/16 OD x 0 125 mm ID x 400 mm length PEEKsil tubing green PK2