Learn More
Trajan Scientific Tubing PEEKSil Green Medical 2/PK
SDP
Catalog No. 5000493671
Encompass_Preferred
1/16 OD x 0 125 mm ID x 150 mm length PEEKsil tubing green PK2
Catalog No. 50-004-93671 Supplier Trajan Scientific Supplier No. 14011001500902
1/16 OD x 0 125 mm ID x 150 mm length PEEKsil tubing green PK2