Learn More
Trajan Scientific Tubing PEEKSil Green Medical 2/PK
SDP
Catalog No. 5000494611
Encompass_Preferred
1/32 OD x 0 125 mm ID x 350 mm length PEEKsil tubing green PK2
Catalog No. 50-004-94611 Supplier Trajan Scientific Supplier No. 14021003500902
1/32 OD x 0 125 mm ID x 350 mm length PEEKsil tubing green PK2