Learn More
Trajan Scientific Tubing PEEKSil Green Medical 2/PK
SDP
Catalog No. 5000494022
Encompass_Preferred
1/16 OD x 0 125 mm ID x 300 mm length PEEKsil tubing green PK2
Catalog No. 50-004-94022 Supplier Trajan Scientific Supplier No. 14011003000902
1/16 OD x 0 125 mm ID x 300 mm length PEEKsil tubing green PK2