Learn More
Trajan Scientific Tubing PEEKSil Grey Medical 2/PK
SDP
Catalog No. 5000493893
Encompass_Preferred
1/16 OD x 0 3 mm ID x 650 mm length PEEKsil tubing gray PK2
Catalog No. 50-004-93893 Supplier Trajan Scientific Supplier No. 14010806500802
1/16 OD x 0 3 mm ID x 650 mm length PEEKsil tubing gray PK2