Learn More
Trajan Scientific Tubing PEEKSil Grey Medical 2/PK
SDP
Catalog No. 5000494060
Encompass_Preferred
1/16 OD x 0 3 mm ID x 500 mm length PEEKsil tubing gray PK2
Catalog No. 50-004-94060 Supplier Trajan Scientific Supplier No. 14010805000802
1/16 OD x 0 3 mm ID x 500 mm length PEEKsil tubing gray PK2