Learn More
Trajan Scientific Tubing PEEKSil Grey Medical 2/PK
SDP
Catalog No. 5000494062
Encompass_Preferred
1/16 OD x 0 3 mm ID x 400 mm length PEEKsil tubing gray PK2
Catalog No. 50-004-94062 Supplier Trajan Scientific Supplier No. 14010804000802
1/16 OD x 0 3 mm ID x 400 mm length PEEKsil tubing gray PK2