Learn More
Trajan Scientific Tubing PEEKSil Grey Medical 2/PK
SDP
Catalog No. 5000494663
Encompass_Preferred
1/16 OD x 0 3 mm ID x 50 mm length PEEKsil tubing gray PK2
Catalog No. 50-004-94663 Supplier Trajan Scientific Supplier No. 14010800500802
1/16 OD x 0 3 mm ID x 50 mm length PEEKsil tubing gray PK2