Learn More
Trajan Scientific Tubing PEEKSil Grey Medical 2/PK
SDP
Catalog No. 5000494666
Encompass_Preferred
1/16 OD x 0 3 mm ID x 300 mm length PEEKsil tubing gray PK2
Catalog No. 50-004-94666 Supplier Trajan Scientific Supplier No. 14010803000802
1/16 OD x 0 3 mm ID x 300 mm length PEEKsil tubing gray PK2