Learn More
Trajan Scientific Tubing PEEKSil Grey Medical 2/PK
SDP
Catalog No. 5000493863
Encompass_Preferred
1/16 OD x 0 3 mm ID x 150 mm length PEEKsil tubing gray PK2
Catalog No. 50-004-93863 Supplier Trajan Scientific Supplier No. 14010801500802
1/16 OD x 0 3 mm ID x 150 mm length PEEKsil tubing gray PK2