Learn More
Trajan Scientific Tubing PEEKSil Grey Medical 2/PK
SDP
Catalog No. 5000493740
Encompass_Preferred
1/16 OD x 0 3 mm ID x 200 mm length PEEKsil tubing gray PK2
Catalog No. 50-004-93740 Supplier Trajan Scientific Supplier No. 14010802000802
1/16 OD x 0 3 mm ID x 200 mm length PEEKsil tubing gray PK2