Learn More
Trajan Scientific Tubing PEEKSil Natural Medical 1/EA
SDP
Catalog No. 5000493953
Encompass_Preferred
1/16 OD x 0 22 mm ID x 260 mm length PEEKSil sample loop
Catalog No. 50-004-93953 Supplier Trajan Scientific Supplier No. 0650010
1/16 OD x 0 22 mm ID x 260 mm length PEEKSil sample loop