Learn More
Trajan Scientific Tubing PEEKSil Orange Medical 2/PK
SDP
Catalog No. 5000494465
Encompass_Preferred
1/16 OD x 0 025 mm ID x 550 mm length PEEKsil tubing orange PK2
Catalog No. 50-004-94465 Supplier Trajan Scientific Supplier No. 14010105500102
1/16 OD x 0 025 mm ID x 550 mm length PEEKsil tubing orange PK2