Learn More
Trajan Scientific Tubing PEEKSil Orange Medical 2/PK
SDP
Catalog No. 5000494036
Encompass_Preferred
360 m OD x 0 025 mm ID x 200 mm length PEEKsil tubing orange PK2
Catalog No. 50-004-94036 Supplier Trajan Scientific Supplier No. 14030102000102
360 m OD x 0 025 mm ID x 200 mm length PEEKsil tubing orange PK2