Learn More
Trajan Scientific Tubing PEEKSil Orange Medical 2/PK
SDP
Catalog No. 5000493866
Encompass_Preferred
1/32 OD x 0 025 mm ID x 400 mm length PEEKsil tubing orange PK2
Catalog No. 50-004-93866 Supplier Trajan Scientific Supplier No. 14020104000102
1/32 OD x 0 025 mm ID x 400 mm length PEEKsil tubing orange PK2