Learn More
Trajan Scientific Tubing PEEKSil Orange Medical 2/PK
SDP
Catalog No. 5000493743
Encompass_Preferred
1/32 OD x 0 025 mm ID x 100 mm length PEEKsil tubing orange PK2
Catalog No. 50-004-93743 Supplier Trajan Scientific Supplier No. 14020101000102
1/32 OD x 0 025 mm ID x 100 mm length PEEKsil tubing orange PK2