Learn More
Trajan Scientific Tubing PEEKSil Orange Medical 2/PK
SDP
Catalog No. 5000494071
Encompass_Preferred
1/32 OD x 0 025 mm ID x 750 mm length PEEKsil tubing orange PK2
Catalog No. 50-004-94071 Supplier Trajan Scientific Supplier No. 14020107500102
1/32 OD x 0 025 mm ID x 750 mm length PEEKsil tubing orange PK2