Learn More
Trajan Scientific Tubing PEEKSil Orange Medical 2/PK
SDP
Catalog No. 5000494297
Encompass_Preferred
1/16 OD x 0 025 mm ID x 500 mm length PEEKsil tubing orange PK2
Catalog No. 50-004-94297 Supplier Trajan Scientific Supplier No. 14010105000102
1/16 OD x 0 025 mm ID x 500 mm length PEEKsil tubing orange PK2