Learn More
Trajan Scientific Tubing PEEKSil Orange Medical 2/PK
SDP
Catalog No. 5000493899
Encompass_Preferred
1/32 OD x 0 025 mm ID x 150 mm length PEEKsil tubing orange PK2
Catalog No. 50-004-93899 Supplier Trajan Scientific Supplier No. 14020101500102
1/32 OD x 0 025 mm ID x 150 mm length PEEKsil tubing orange PK2