Learn More
Trajan Scientific Tubing PEEKSil Purple Medical 2/PK
SDP
Catalog No. 5000493916
Encompass_Preferred
1/16 OD x 0 15 mm ID x 550 mm length PEEKsil tubing purple PK2
Catalog No. 50-004-93916 Supplier Trajan Scientific Supplier No. 14010505500502
1/16 OD x 0 15 mm ID x 550 mm length PEEKsil tubing purple PK2