Learn More
Trajan Scientific Tubing PEEKSil Purple Medical 2/PK
SDP
Catalog No. 5000494063
Encompass_Preferred
1/16 OD x 0 15 mm ID x 700 mm length PEEKsil tubing purple PK2
Catalog No. 50-004-94063 Supplier Trajan Scientific Supplier No. 14010507000502
1/16 OD x 0 15 mm ID x 700 mm length PEEKsil tubing purple PK2